Hematopoietic Progenitor Cell Medium, PromoCell
Supplier: Promocell, Inc.
C-28021
10172-388EA 317.86 USD
10172-388
Hematopoietic Progenitor Cell Medium, PromoCell
Cell Culture Media
The PromoCell Hematopoietic Progenitor Cell (HPC) Expansion Medium DXF has been designed for the maximum expansion of primitive HPCs in a fully defined, xeno-free culture environment
The PromoCell Hematopoietic Progenitor Medium has been developed for the expansion of hematopoietic progenitors by means of differentiation into more committed lineages of blood cells. Hence, this progenitor medium is suitable for CFU-based assays, e.g. long-term culture initiating cell (LTC-IC) assays.
Recommended for Human Mononuclear Cells (hMNC), Human CD34+ Progenitor Cells (hCD34+-CB), Human CD133+ Progenitor Cells (hCD133+-CB), Human Hematopoietic Progenitor Cells (HPCs)
Due to the utilization of exclusively synthetic, recombinant or plant-sourced materials, this medium is chemically defined and free of all animal-derived components and substances of human origin with Human Serum Albumin as the only exception. Note: The medium must be supplemented with additional cytokines, e.g. Cytokine Mix E (see below). Individual cytokines can be found on the PromoKine website.
Ordering information: The Hematopoietic Progenitor Expansion Medium DXF consists of a bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Medium. In order to achieve optimal performance, the complete Medium must be supplemented with additional growth factors by the user. PromoCell Hematopoietic Progenitor Media contain all the growth factors and supplements necessary for the efficient expansion or directed differentiation of blood-derived progenitors into hematopoietic lineages.