Osteoblast Mineralization Medium, PromoCell
Supplier: Promocell, Inc.
C-27020
10172-344EA 272.09 USD
10172-344
Osteoblast Mineralization Medium, PromoCell
Cell Culture Media
PromoCell Osteoblast Mineralization Medium was developed for the efficient induction of strong mineralization in human osteoblast cultures
Extensive extracellular calcium-deposits can be observed after 21 days, following an easy one-step protocol.