Anti-IgG (H+L) Rabbit Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Southern Biotechnology
6160-02
100245-142EA 91.54 USD
100245-142
Anti-IgG (H+L) Rabbit Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Rabbit Anti-Goat IgG(H+L)-FITC antibody for use in immunohistochemistry / immunocytochemistry and western blotting assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR