Anti-IgG (H+L) Rabbit Antibody

Supplier: Southern Biotechnology
6150-01
100245-118EA 75.85 USD
100245-118
Anti-IgG (H+L) Rabbit Antibody
Antibodies
Purified Rabbit Anti-Sheep IgG(H+L) antibody for use in ELISA, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR