Anti-Kappa Mouse Antibody (FITC (Fluorescein Isothiocyanate)) [clone: K4F5]

Supplier: Southern Biotechnology
3090-02
100244-640EA 499.54 USD
100244-640
Anti-Kappa Mouse Antibody (FITC (Fluorescein Isothiocyanate)) [clone: K4F5]
Antibodies
FITC Anti-Rat Kappa antibody for use in flow cytometry, ELISA, immunohistochemistry, and western blotting assays.

Type: Secondary
Antigen: Kappa
Clonality: Monoclonal
Clone: K4F5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR