Anti-CD11b Rat Monoclonal Antibody (Cy5®) [clone: 3A33]

Supplier: Southern Biotechnology
1560-15
100243-222EA 377.92 USD
100243-222
Anti-CD11b Rat Monoclonal Antibody (Cy5®) [clone: 3A33]
Antibodies
CD11b (integrin αM subunit) combines with CD18 (integrin β2 subunit) to form the integrin Mac-1, also known as complement receptor 3 (CR3), which mediates adhesion to C3bi and ICAM-1 (CD54). Mac-1 is expressed at varying levels on granulocytes, macrophages, dendritic cells, NK cells, and B-1 cells in the peritoneal and pleural cavities. In addition to its role in binding C3bi on opsonized targets and mediation of the subsequent ingestion process, Mac-1 is important as an adhesion molecule in the transendothelial migration of monocytes and neutrophils.

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: 3A33
Conjugation: Cy5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR