Anti-CD3 epsilon Hamster Monoclonal Antibody [clone: 500-A2]

Supplier: Southern Biotechnology
1531-14
100243-088EA 299.46 USD
100243-088
Anti-CD3 epsilon Hamster Monoclonal Antibody [clone: 500-A2]
Antibodies
CD3ε, a member of the immunoglobulin superfamily of cell surface receptors, is comprised of five invariable chains ranging in size from 16-28 kDa and is closely associated with the T cell antigen receptor (TCR). CD3ε is expressed on all T cells of all mouse strains. CD3 plays a major role in signaling during antigen recognition, leading to T-cell activation. 500-A2 recognizes an epitope on the 25 kDa ε chain of the CD3/TCR complex of mouse strains tested.

Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: 500-A2
Conjugation:
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR