Anti-Type III Collagen Goat Polyclonal Antibody (Biotin)

Supplier: Southern Biotechnology
1330-08
100242-972EA 589.77 USD
100242-972
Anti-Type III Collagen Goat Polyclonal Antibody (Biotin)
Antibodies
Biotin Anti-Type III Collagen antibody for use in ELISA, immunohistochemistry / immunocytochemistry, electron microscopy, flow cytometry, and western blotting assays.

Type: Primary
Antigen: Type III Collagen
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Canine, Chicken, Chinchilla, Equine, Guinea Pig, Hamster, Porcine, Rabbit, Rhesus, Tree Shrew
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR