Anti-IgG (H+L) Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
1032-05
100242-526EA 200.08 USD
100242-526
Anti-IgG (H+L) Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat F(ab')2 Anti-Mouse IgG(H+L)-HRP antibody with minimal reactivity to human proteins for use in ELISA, immunohistochemistry / immunocytochemistry, western blotting, and ELISpot assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR