Anti-IL-4 Rat Monoclonal Antibody [clone: BVD6-24G2]

Supplier: Southern Biotechnology
10204-01
100242-396EA 333.46 USD
100242-396
Anti-IL-4 Rat Monoclonal Antibody [clone: BVD6-24G2]
Antibodies
Purified Anti-Mouse IL-4 antibody for use in flow cytometry, immunohistochemistry, western blotting, immunoprecipitation, ELISA, and ELISpot assays.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: BVD6-24G2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR