Mouse IgG Isotype Control (Spectral Red® RPE (R-Phycoerythrin)/Cy5®)

Supplier: Southern Biotechnology
0107-13
100241-996EA 241.92 USD
100241-996
Mouse IgG Isotype Control (Spectral Red® RPE (R-Phycoerythrin)/Cy5®)
Antibodies
Mouse IgG-PE/CY5 (SPRD) for use as a control in flow cytometry assays.

Type: Control
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Spectral Red® RPE (R-Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR