Mouse IgG3 Isotype Control [clone: B10] (FITC (Fluorescein Isothiocyanate))

Supplier: Southern Biotechnology
0105-02
100241-954EA 197.46 USD
100241-954
Mouse IgG3 Isotype Control [clone: B10] (FITC (Fluorescein Isothiocyanate))
Antibodies
Mouse IgG3-FITC for use as a control in flow cytometry and immunocytochemistry assays.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: B10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR