Introducing our New VWR Standard Tissue Culture Line!
![](/m-us.vwr.com/en_US/images/vwr_tissue_culture_200.jpg)
We understand the need to stretch your dollars and the need for quality. Our New VWR Collection Tissue Culture Consumables can help you do both!
These items are vacuum-gas and plasma treated to ensure consistent cell attachment, spreading and growth.
- Manufactured in a Class 100 cleanroom environment
- Produced with 100% USP VI crystal virgin polystyrene
- Sterilized by gamma irradiation
- Certified non-pyrogenic
Providing a great value for routine tissue culture!