sparQ Next Generation Sequencing (NGS) Solutions
sparQ Next Generation Sequencing (NGS) Solutions
Quantabio now offers a full suite of sparQ NGS DNA products that provide streamlined workflows solutions with improved sequencing performance for library fragmentation, preparation, amplification, and quantification for sequencing on Illumina® NGS platforms.
sparQ NGS DNA products enable:
- reduced process time and hands-on time
- maximized library yields
- improved coverage uniformity
- minimized bias
- reliable and reproducible results
Product selection
sparQ DNA Frag & Library Prep
sparQ DNA Frag and Library Prep Kit optimizes the integration of enzymatic fragmentation into a two-step protocol for the streamlined construction of libraries for sequencing on Illumina® NGS platforms.
sparQ HiFi PCR Master Mix
A high-efficiency, high-fidelity, and low-bias PCR master mix for NGS library amplification.
sparQ DNA Library Prep
sparQ DNA Library Prep Kit is an optimized kit for rapid construction of DNA libraries from fragmented double-stranded DNA for sequencing on Illumina NGS platforms.
sparQ PureMag Beads
A fast and reliable nucleic acid purification system for reaction cleanup and size selection in Next Generation Sequencing (NGS) workflows.
sparQ Universal Library Quant Kit
Rapid and accurate Real-Time qPCR kit for quantifying NGS Library molecules prior to sequencing