Laser Microdissection
![](/m-us.vwr.com/en_US/images/csm_move-the-laser_15_89be1421d0_200x200.jpg)
Laser Microdissection (LMD, also known as Laser Capture Microdissection or LCM) enables users to isolate specific single cells or entire areas of tissue. Powered by a unique laser design and dynamic software, Leica LMD systems allow users to easily isolate Regions of Interest (ROI) from entire areas of tissue down to single cells or even subcellular structures such as chromosomes.
Product selection
![](/m-us.vwr.com/en_US/images/LMD7_DFC7000_15_100x100.jpg)
Laser Microdissection
Laser Microdissection (LMD) enables users to isolate specific single cells or entire areas of tissue for downstream genomic and proteomic analyses.