Industrial Gloves
![](/m-us.vwr.com/en_US/images/30848_APP_Y9296_300_duct_w_sleve-child-3.jpg)
Wells Lamont products can help protect your hands in many industrial situations. Options include cut resistant, leather, mechanics, impact, heat resistant, palm dipped, and general purpose. We not only support critical environment applications but also maintenance, food processing, manufacturing, metal handling, welding, metal stamping, shipping, oil and gas, and many more.
![](/m-us.vwr.com/en_US/images/30848_Leather_Glove-child-3.jpg)
Leather Glove
Our Y0122 is a high quality grain cowhide drivers glove that offers high abrasion and puncture resistance.
![](/m-us.vwr.com/en_US/images/30848_Metal_Mesh_Glove-child-3.jpg)
Metal Mesh Glove
This glove offers superior cut resistance and features a patented self tensioning cuff that is easier to put on and more hygenic.
![](/m-us.vwr.com/en_US/images/30848Heat_Resistance_up_to_350_F-child-3.jpg)
Heat Resistance up to 350 F
The terry cloth 636HR offers excellent heat resistance up to 350F and features a 5" gauntlet for protecting the wrist area.
![](/m-us.vwr.com/en_US/images/30848_Palm_Dipped_Glove-child-3.jpg)
Palm Dipped Glove
The Y9265 has a polyurethane palm for grip and offers ANSI A2 cut resistance.
![](/m-us.vwr.com/en_US/images/30848_Sleeve_with_ANSI_A7_cut_resistance-child-3.jpg)
Sleeve with ANSI A7 cut resistance
The Armguard II Medium Duty is a 20" high level cut resistant sleeve that is made in the USA.