Weighing Boats & Dishes
From microgram quantities to larger samples, find the perfect fit for your lab weighing needs with our extensive selection of weighing boats & dishes.
Standard Weighing Boats
These flexible polystyrene weighing boats are disposable and feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Aluminum Weighing Dishes
Great for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.
Pour-Boat Weighing Dishes
Designed to easily dispense samples without flexing the dish. Perfect for easy sample handling.
Sterile Weighing Boats
Ideal for applications where there is concern of cross contamination and ‘clean’ is a necessity in life science research, food, bio-pharmaceutical, environmental and molecular research environments.