Crusher/Splitter Hoods
![](/m-us.vwr.com/en_US/images/107239-14L_200x200_main.jpg)
The crusher/splitter hoods effectively capture and collect dust while providing uniform airflow, a clean work surface and protecting the operator from contaminants. Hood can have the option to have your crusher placed on the left- or right-hand side to provide a convenient work area for a sample splitter. Designed mining applications, our crusher/splitter hoods have a 10” flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector (sold separately, contact us for more information).
Crusher/splitter hoods are made of steel and blue powder coated for a long-lasting finish (additional colours available on request). Each unit comes complete with LED lighting and a compressed air hose. Custom sizes are also available.
- Splitter Hood 5’ Left Hand - CA11022-178 (Supplier number 107239-15L)
- Splitter Hood 5’ Right Hand - CA11022-180 (Supplier number 107239-15R)
- Crusher/splitter hood Left Hand - CA11022-174 (Supplier number 107239-14L)
- Crusher/splitter hood Right Hand - CA11022-176 (Supplier number 107239-14R)
- Crusher/RSD hood - CA11022-182 (Supplier number 107239-16)