Azure cSeries Advanced Imaging Systems
c600 | c500 | c400 | c300 |
---|---|---|---|
Ultimate Western SystemIR Fluorescence |
Infrared Imaging SystemIR Fluorescence Upgradeable to c600 |
Visible Fluorescent |
Darkroom EliminatorChemiluminescence Upgradeable to c600, c500, and c400 |
![]() |
![]() |
![]() |
![]() |
cSeries Specifications | c600 | c500 | c400 | c300 |
---|---|---|---|---|
Camera | 8.3 MP | |||
Cooling | -50°C regulated cooling | |||
7 Position Filter Wheel | ✓ | ✓ | ✓ | ✓ |
UV 302/365 nm | ✓ | ✓ | ✓ | ✓ |
EPI Blue LED | ✓ | ✓ | ✓ | ✓ |
Chemi | ✓ | ✓ | ✓ | ✓ |
Visible Fluorescent Westerns | ✓ | ✓ | ||
NIR Westerns | ✓ | ✓ | ||
Field of View | 20 × 15 cm | |||
Footprint (W × H × D) | 38 × 55 × 36 cm |