IG® sgRNA Synthesis Kits for Cas9 Nuclease
Supplier: INTACT GENOMICS, INC MS
3203 3206
104090-504PK 252.69 USD
104090-504 104090-506
IG® sgRNA Synthesis Kits for Cas9 Nuclease
Enzymes
Intact Genomics is your resource for in vitro gene editing using Cas9 enzymes. We understand your need to generate sufficient quantities of sgRNA for in vitro gene editing projects. The IG® sgRNA synthesis kits are simple to use, and our kits are scalable to help you reach these goals. The IG® sgRNA synthesis kits can be purchased with or without the sgRNA purification kit.
  • IG® sgRNA Synthesis Kits are a perfect choice for a variety of sgRNA synthesis needs
  • IG® sgRNA Synthesis Kit includes everything but your targeting oligo (all buffers, enzymes, scaffolds)
  • User simply needs to provide the oligo for your target - must be 55 base pairs and generated using the template design in this manual
  • Rapid workflow (Less than 1 hour)
  • Scalable yield (just 1 reaction for all the sgRNA you need)
With high and scalable yield, the IG® sgRNA synthesis kit for Cas9 nuclease is the perfect choice for a variety of sgRNA synthesis needs.
Shipping temperature: Dry ice
Certifications:
Manufactured under ISO 13485:2016 Quality Management certified standards.