K599 electrocompetent Agrobacterium cells
Supplier: INTACT GENOMICS, INC MS
1287-36 1287-12
104090-476PK 865.38 USD
104090-476 104090-474
K599 electrocompetent Agrobacterium cells
Competent Cells
Made from Agrobacterium rhizogenes (str R) pRi2659 (agropine type).
  • Useful for transgenic operations of corn, soybean (wild soybean), cotton, peanut, dandelion, cowpea and other plants.
  • Transformation efficiency:  ≥ 1×104  cfu/µg with control plasmid
Intact Genomics (ig®) K599 Agrobacterium Chemically Competent Cells are made from a specific strain of Agrobacterium rhizogenes, Agrobacterium rhizogenes (str R) pRi2659 (agropine type). K599 was originally isolated from cucumber exhibiting hairy root disease symptoms and has been widely used for hairy root transformation.
Agrobacterium rhizogenes is a soil-borne gram-negative bacterium that can infect most dicotyledons, a few monocotyledons and some gymnosperms.  K599 Chemically Competent Agrobacterium are optimized for the highest transformation efficiencies and are useful for transgenic operations of corn, soybean (wild soybean), cotton, peanut, dandelion, cowpea and other plants.  K599 Agrobacterium rhizogenes strain contains pRi2659 agrobacterium-type Ri plasmid and displays streptomycin resistance.
Certifications:
Manufactured under ISO 13485:2016 certified quality management system