ig® Methionine Auxotrophic EHA105ᴹᵉᵗ Chemically Competent Agrobacterium, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
1078-05 1078-15
104090-450PK 259.62 USD
104090-450 104090-452
ig® Methionine Auxotrophic EHA105ᴹᵉᵗ Chemically Competent Agrobacterium, Intact Genomics
Competent Cells
Methionine Auxotrophic EHA105ᴹᵉᵗ Chemically Competent Agrobacterium.
  • Optimized for the highest transformation efficiencies
  • Requires supplemental methionine for growth
  • Useful for transgenic operations of plants
  • Amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA)
Intact Genomics (ig®) Methionine-Auxotrophic EHA105Met Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless methionine is added to minimal medium.
This prevents the bacteria from overgrowing plant tissues when used for plant transformation.
Methionine- Auxotrophic EHA105Met Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies.
The EHA105Met strain is useful for transgenic operations of rice, tobacco and other plants. EHA105 contains a rifampicin resistance gene (rif). EHA105Met strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function, which contains the vir gene.
Certifications:
Manufactured under ISO 13485:2016 certified quality management system.