Human Recombinant BMP-4 (from E. coli)
Supplier: VWR International
77751-116 77751-118 77751-120 77751-122 77751-124
77751-116EA 2684.08 USD
77751-116 77751-118 77751-120 77751-122 77751-124
Human Recombinant BMP-4 (from E. coli)
Proteins and Peptides
Bone morphogenetic protein 4 (BMP-4) is a member of the bone morphogenetic sub-family (BMP) and functions as a critical element involved in the culture and differentiation of stem cells. Active BMP-4 consists of forming a homodimer or a heterodimer with related BMPs, such as BMP-7. BMP-4 is integral in stem cell cultures and the differentiation of mesoderm and ectoderm from pluripotent stem cells (iPSCs and ESCs). BMP-4 is used in protocols for culturing cardiomyocytes, early hematopoietic cells, hepatocytes, bone, cartilage, adipocytes and lung, as well as, inhibition of early neuronal differentiation.
  • High quality
  • Low endotoxin
  • Affordable
Caution:
For research use only.