Brady® i75 R4500 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176790 176791 176792 176793 176789
77775-096RL 0 USD
77775-096 77775-098 77775-100 77775-102 77775-094
Brady® i75 R4500 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbin ink wind direction: ink side out
  • Polyester material
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R4500 Series colored wax/resin formulation offers smear resistance and excellent barcode read rates when printed on the appropriate label material from Brady®.
Recommended for printing on paper and films that have a matte or rough surface, such as B-427 Self-Laminating Vinyl and B-499 Nylon Cloth from Brady®. Combine Brady label materials with their recommended ribbon series for superior legend performance.