NEBNext® RSV Primer Modules
Supplier: New England Biolabs (NEB)
E9642L
E9642S
77868-076EA
1151.07
USD
77868-076
77868-078
NEBNext® RSV Primer Modules
Nucleic Acid Reagents
Primers and Probes
The NEBNext® RSV Primer Module contains the primers required for RT-PCR upstream of library prep for sequencing on Illumina or Oxford Nanopore Technologies platforms.
Curated primer mix for full-length sequencing
- Covers RSV A and RSV B with a single workflow
- Faster answers than targeted panels
- Appropriate for variant analysis
- Compatible with NEBNext® Primer Monitor tool
The module includes primers for full-length sequencing of both RSV A and RSV B.
Caution:
The module is intended for research use only.