Chart Recorders for Ultra-Low Temperature Freezers
Supplier: Thermo Fisher Scientific
6383-6A 6383-6D 6383-6V
14228-376EA 2220 USD
14228-376 77681-926 77681-928
Chart Recorders for Ultra-Low Temperature Freezers
Freezers Ultra Low Temperature Freezers
External chart recorders provide independent temperature recording and record keeping for your ultra-low temperature freezers.
  • Stand-alone, independent probe
  • Seven day circular chart recorder
  • Use 15 cm (6") chart paper
  • Functions to −150 °C
Certifications:
CE.