United Scientific Digital Pocket Scales
Supplier: United Scientific Supplies
UNEDSCL-600 UNEDSCL-100 UNEDSCL-200
470356-486EA 14.43 USD
470356-486 470356-524 470356-526
United Scientific Digital Pocket Scales
Balances and Scales
Pocket size power for measuring weight.
  • Four weighing modes
  • Auto shut-off after 60 seconds
These compact scales have a 55×55 mm weighing platform. Each features four weighing modes: g/oz/ozt/dwt, tare function, and a 60 second auto shut-off to preserve battery life. Available in three different capacities. All use 2 'AAA' batteries (included).
Delivery information:
Includes a dual-purpose cover/weighing tray.