Prostaglandin E2 ELISA Kit - Extracellular
Supplier: Abcam
AB316263-96TESTS AB316263-10X96TESTS
ABCA_AB316263-96TEEA 1009 USD
ABCA_AB316263-96TE ABCA_AB316263-10X9
Prostaglandin E2 ELISA Kit - Extracellular
Assays ELISAs
Prostaglandin E2 ELISA Kit - Extracellular is a single-wash 90-min Simplestep used to quantify Prostaglandin E2 with a sensitivity of 4.75 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Competitive ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.