HIV1 p24 ELISA Kit- Extracellular
Supplier: Abcam
AB320051-10X96TESTS AB320051-1X384TESTS AB320051-1X96TESTS
ABCA_AB320051-10X9EA 7702 USD
ABCA_AB320051-10X9 ABCA_AB320051-1X38 ABCA_AB320051-1X96
HIV1 p24 ELISA Kit- Extracellular
Assays ELISAs
HIV1 p24 ELISA Kit- Extracellular is a single-wash 90-min Simplestep used to quantify HIV1 p24 with a sensitivity of 4.3 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput