Human IL-2 ELISA Set (without plates)
Supplier: Abcam
AB48471-15X96TESTS AB48471-5X96TESTS AB48471-10X96TESTS
ABCA_AB48471-15X96EA 1466 USD
ABCA_AB48471-15X96 ABCA_AB48471-5X96T ABCA_AB48471-10X96
Human IL-2 ELISA Set (without plates)
Assays ELISAs
Human IL-2 ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-2 ELISA Set (without plates) in Human in Plasma, Cell culture supernatant, Serum samples.