Mouse IgE ELISA Kit- Extracellular
Supplier: Abcam
AB317773-1X384TESTS AB317773-10X96TESTS AB317773-1X96TESTS
ABCA_AB317773-1X38EA 2388 USD
ABCA_AB317773-1X38 ABCA_AB317773-10X9 ABCA_AB317773-1X96
Mouse IgE ELISA Kit- Extracellular
Assays ELISAs
Mouse IgE ELISA Kit- Extracellular is a single-wash 90-min Simplestep used to quantify Mouse IgE with a sensitivity of 7.263 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput .