Human IFN gamma High Sensitivity ELISA Kit
Supplier: Abcam
AB46048-2X96TESTS AB46048-1X96TESTS
ABCA_AB46048-2X96TEA 1448 USD
ABCA_AB46048-2X96T ABCA_AB46048-1X96T
Human IFN gamma High Sensitivity ELISA Kit
Assays ELISAs
Human IFN gamma High Sensitivity ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IFN gamma High Sensitivity in Human in Plasma, Cell culture supernatant, Serum samples.