HIV1 p24 ELISA Kit- Intracellular
Supplier: Abcam
AB320057-1X96TESTS AB320057-10X96TESTS AB320057-1X384TESTS
ABCA_AB320057-1X96EA 859 USD
ABCA_AB320057-1X96 ABCA_AB320057-10X9 ABCA_AB320057-1X38
HIV1 p24 ELISA Kit- Intracellular
Assays ELISAs
HIV1 p24 ELISA kit- Intracellular is a single-wash 90-min Simplestep used to quantify HIV1 p24 with a sensitivity of 4.8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.