Human IL-8 ELISA Kit
Supplier: Abcam
AB214030-1X384TESTS AB214030-10X96TESTS AB214030-1X96TESTS
ABCA_AB214030-1X38EA 2388 USD
ABCA_AB214030-1X38 ABCA_AB214030-10X9 ABCA_AB214030-1X96
Human IL-8 ELISA Kit
Assays ELISAs
Human IL-8 ELISA kit is a single-wash, 90-minute SimpleStep ELISA® used to quantify human interleukin-8 (IL-8) with a sensitivity of 1.8 pg/mL. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450 nm readout; compatible with any standard plate reader - Available in different formats to meet various throughput needs, including 384-well plates for higher throughput - Broad sample compatibility – suitable for serum, plasma, cell culture supernatants, and cerebrospinal fluid - Cited in over 30 publications