Human IDO ELISA Kit
Supplier: Abcam
AB245710-10X96TESTS AB245710-1X384TESTS AB245710-1X96TESTS
ABCA_AB245710-10X9EA 8335 USD
ABCA_AB245710-10X9 ABCA_AB245710-1X38 ABCA_AB245710-1X96
Human IDO ELISA Kit
Human IDO ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IDO with a sensitivity of 13 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.