Human IL-2 ELISA Kit
Supplier: Abcam
AB270883-10X96TESTS AB270883-1X384TESTS AB270883-1X96TESTS
ABCA_AB270883-10X9EA 8460 USD
ABCA_AB270883-10X9 ABCA_AB270883-1X38 ABCA_AB270883-1X96
Human IL-2 ELISA Kit
Assays ELISAs
Human IL-2 ELISA Kit is a single-wash, 90-minute SimpleStep ELISA® used to quantify human interleukin-2 (IL-2) with a sensitivity of 32.1 pg/ml. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric Sandwich ELISA - 450 nm readout; compatible with any standard plate reader - Available in different formats to meet various throughput needs, including 384-well plates for higher throughput - Broad sample compatibility – suitable for cell culture media, cell culture supernatant, serum and plasma.