Human EGF ELISA Kits
Supplier: Abcam
AB217772-1X96TESTS AB217772-1X384TESTS AB217772-10X96TESTS
ABCA_AB217772-1X96EA 1009 USD
ABCA_AB217772-1X96 ABCA_AB217772-1X38 ABCA_AB217772-10X9
Human EGF ELISA Kits
Assays ELISAs
Human EGF ELISA Kit is a single-wash 90-min Simplestep used to quantify Human EGF with a sensitivity of 1.04 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput. - Cited in over 5 citations.