Mouse Interferon gamma ELISA Set (without plates)
Supplier: Abcam
AB47618-10X96TESTS AB47618-5X96TESTS AB47618-15X96TESTS
ABCA_AB47618-10X96EA 1228 USD
ABCA_AB47618-10X96 ABCA_AB47618-5X96T ABCA_AB47618-15X96
Mouse Interferon gamma ELISA Set (without plates)
Assays ELISAs
Mouse Interferon gamma ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Interferon gamma ELISA Set (without plates) in Mouse in plasma, cell culture supernatant, serum samples.