Anti-RNA polymerase beta Rabbit Monoclonal Antibody [clone: EPR18704]

Supplier: Abcam
AB191598-100UL AB191598-1ML
ABCA_AB191598-100UEA 652 USD
ABCA_AB191598-100U ABCA_AB191598-1ML
Anti-RNA polymerase beta Rabbit Monoclonal Antibody [clone: EPR18704]
Antibodies
Rabbit monoclonal [EPR18704] to RNA polymerase beta.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RNA polymerase beta
Clonality: Monoclonal
Clone: EPR18704
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR