Anti-SMC1A Rabbit Monoclonal Antibody [clone: EP2879Y]

Supplier: Abcam
AB75819-100UL AB75819-1ML
ABCA_AB75819-100ULEA 677 USD
ABCA_AB75819-100UL ABCA_AB75819-1ML
Anti-SMC1A Rabbit Monoclonal Antibody [clone: EP2879Y]
Antibodies
Rabbit monoclonal [EP2879Y] to SMC1A.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SMC1A
Clonality: Monoclonal
Clone: EP2879Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR