Anti-RNA polymerase II CTD repeat YSPTSPS Rat Monoclonal Antibody [clone: 1C7]

Supplier: Abcam
AB255848-100UG AB255848-1MG
ABCA_AB255848-100UEA 1134 USD
ABCA_AB255848-100U ABCA_AB255848-1MG
Anti-RNA polymerase II CTD repeat YSPTSPS Rat Monoclonal Antibody [clone: 1C7]
Antibodies
Rat monoclonal [1C7] to RNA polymerase II CTD repeat YSPTSPS - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: RNA polymerase II CTD repeat YSPTSPS
Clonality: Monoclonal
Clone: 1C7
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR