Anti-NDRG1 Rabbit Monoclonal Antibody [clone: EPR5593]

Supplier: Abcam
AB216457-1MG AB216457-100UG
ABCA_AB216457-1MGEA 10227 USD
ABCA_AB216457-1MG ABCA_AB216457-100U
Anti-NDRG1 Rabbit Monoclonal Antibody [clone: EPR5593]
Antibodies
Rabbit monoclonal [EPR5593] to NDRG1 - Low endotoxin, Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: NDRG1
Clonality: Monoclonal
Clone: EPR5593
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR