Anti-CRISPR-Cas9 Rat Monoclonal Antibody [clone: KANI345B]

Supplier: Abcam
AB271293-1MG AB271293-100UG
ABCA_AB271293-1MGEA 6856 USD
ABCA_AB271293-1MG ABCA_AB271293-100U
Anti-CRISPR-Cas9 Rat Monoclonal Antibody [clone: KANI345B]
Antibodies
Rat monoclonal [KANI345B] to CRISPR-Cas9.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CRISPR-Cas9
Clonality: Monoclonal
Clone: KANI345B
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR