Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S5) Rabbit Recombinant Multiclonal Antibody [clone: RM1152]

Supplier: Abcam
AB317456-20UL AB317456-1ML AB317456-100UL
ABCA_AB317456-20ULEA 232 USD
ABCA_AB317456-20UL ABCA_AB317456-1ML ABCA_AB317456-100U
Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S5) Rabbit Recombinant Multiclonal Antibody [clone: RM1152]
Antibodies
Rabbit recombinant multiclonal [RM1152] to RNA polymerase II CTD repeat YSPTSPS (phospho S5).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RNA polymerase II CTD repeat YSPTSPS
Clonality: Recombinant Multiclonal
Clone: RM1152
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR