Anti-Rb Rabbit Monoclonal Antibody [clone: EPR17512] (Alexa Fluor® 594)

Supplier: Abcam
AB300670-100UL AB300670-10UL
ABCA_AB300670-100UEA 815 USD
ABCA_AB300670-100U ABCA_AB300670-10UL
Anti-Rb Rabbit Monoclonal Antibody [clone: EPR17512] (Alexa Fluor® 594)
Antibodies
Alexa Fluor® 594 Rabbit monoclonal [EPR17512] to Rb.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Rb
Clonality: Monoclonal
Clone: EPR17512
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR