Anti-Neuroligin 2 Rabbit Recombinant Multiclonal Antibody [clone: RM2048]

Supplier: Abcam
AB317510-1ML AB317510-100UL AB317510-20UL
ABCA_AB317510-1MLEA 5985 USD
ABCA_AB317510-1ML ABCA_AB317510-100U ABCA_AB317510-20UL
Anti-Neuroligin 2 Rabbit Recombinant Multiclonal Antibody [clone: RM2048]
Antibodies
Rabbit recombinant multiclonal [RM2048] to Neuroligin 2.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Neuroligin 2
Clonality: Recombinant Multiclonal
Clone: RM2048
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR