Anti-GluN2C Rabbit Monoclonal Antibody [clone: EPR19046]

Supplier: Abcam
AB192831-100UL AB192831-1ML AB192831-20UL
ABCA_AB192831-100UEA 620 USD
ABCA_AB192831-100U ABCA_AB192831-1ML ABCA_AB192831-20UL
Anti-GluN2C Rabbit Monoclonal Antibody [clone: EPR19046]
Antibodies
Rabbit monoclonal [EPR19046] to GluN2C.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GluN2C
Clonality: Monoclonal
Clone: EPR19046
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR