Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S7) Rat Monoclonal Antibody [clone: 4000000000000]

Supplier: Abcam
AB252853-100UG AB252853-1MG
ABCA_AB252853-100UEA 708 USD
ABCA_AB252853-100U ABCA_AB252853-1MG
Anti-RNA polymerase II CTD repeat YSPTSPS (phospho S7) Rat Monoclonal Antibody [clone: 4000000000000]
Antibodies
Rat monoclonal [4E12] to RNA polymerase II CTD repeat YSPTSPS (phospho S7).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RNA polymerase II CTD repeat YSPTSPS
Clonality: Monoclonal
Clone: 4000000000000
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR