Anti-Myosin light chain kinase/MLCK Rabbit Monoclonal Antibody [clone: EP1458Y]

Supplier: Abcam
AB76092-10UL AB76092-100UL AB76092-1ML
ABCA_AB76092-10ULEA 295 USD
ABCA_AB76092-10UL ABCA_AB76092-100UL ABCA_AB76092-1ML
Anti-Myosin light chain kinase/MLCK Rabbit Monoclonal Antibody [clone: EP1458Y]
Antibodies
Rabbit monoclonal [EP1458Y] to Myosin light chain kinase/MLCK.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Myosin light chain kinase/MLCK
Clonality: Monoclonal
Clone: EP1458Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR