Anti-RNA polymerase II CTD repeat YSPTSPS (phospho T4) Rat Monoclonal Antibody [clone: 6D7]

Supplier: Abcam
AB255845-1MG AB255845-100UG
ABCA_AB255845-1MGEA 10227 USD
ABCA_AB255845-1MG ABCA_AB255845-100U
Anti-RNA polymerase II CTD repeat YSPTSPS (phospho T4) Rat Monoclonal Antibody [clone: 6D7]
Antibodies
Rat monoclonal [6D7] to RNA polymerase II CTD repeat YSPTSPS (phospho T4) - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RNA polymerase II CTD repeat YSPTSPS
Clonality: Monoclonal
Clone: 6D7
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR